Learn More
Medchemexpress LLC 3,6,9,12-Tetraoxatetradecane-1,14-diamine | 68960-97-4 | 98.0% | 236.31 | 250 MG
SDP
Catalog No. 5000403105
Encompass_Preferred
Amino-PEG4-C2-amine is a PEG-based (4 units) PROTAC linker that can be used in the synthesis of PROTACs.
  • Peg-based (4 units) protac linker
  • Used in the synthesis of protacs
Catalog No. 50-004-03105 Supplier Medchemexpress LLC Supplier No. 250MGHY22335
Amino-PEG4-C2-amine is a PEG-based (4 units) PROTAC linker that can be used in the synthesis of PROTACs.
  • Peg-based (4 units) protac linker
  • Used in the synthesis of protacs