Learn More
Medchemexpress LLC 3,6,9,12-Tetraoxatetradecanoic acid, 14-hydroxy- | 70678-95-4 | ≥97.0% | C10H20O7 | 250 MG
SDP
Catalog No. 5000352718
Encompass_Preferred
Hydroxy-PEG4-CH2COOH is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-52718 Supplier Medchemexpress LLC Supplier No. 250MGHY140498
Hydroxy-PEG4-CH2COOH is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins