Learn More
Medchemexpress LLC 3,6,9,12-tetraoxatridecan-1-amine | 85030-56-4 | 98.8% | 207.27 g/mol | C9H21NO4 | 5 G
SDP
Catalog No. 5000204111
Encompass_Preferred
m-PEG4-Amine is a methoxy-terminated polyethylene glycol (PEG4) linker with a terminal primary amine used as a cleavable linker for antibody-drug conjugates (ADCs) and as a PEG-based linker in PROTAC synthesis. The compound has CAS 85030-56-4, molecular weight 207.27 g/mol, and is typically supplied as a colorless to light yellow liquid.
  • Primary amine functional group suitable for conjugation chemistry.
  • Methoxy-terminated PEG4 spacer provides solubility and flexibility.
  • Cleavable linker applications for ADC and PROTAC synthesis.
  • Stable when stored protected from light at recommended temperatures.
  • Colorless to light yellow liquid, easy to handle in solution-phase chemistry.
Catalog No. 50-002-04111 Supplier Medchemexpress LLC Supplier No. HYW0402145G
m-PEG4-Amine is a methoxy-terminated polyethylene glycol (PEG4) linker with a terminal primary amine used as a cleavable linker for antibody-drug conjugates (ADCs) and as a PEG-based linker in PROTAC synthesis. The compound has CAS 85030-56-4, molecular weight 207.27 g/mol, and is typically supplied as a colorless to light yellow liquid.
  • Primary amine functional group suitable for conjugation chemistry.
  • Methoxy-terminated PEG4 spacer provides solubility and flexibility.
  • Cleavable linker applications for ADC and PROTAC synthesis.
  • Stable when stored protected from light at recommended temperatures.
  • Colorless to light yellow liquid, easy to handle in solution-phase chemistry.