Learn More
Medchemexpress LLC 3,6,9,12-tetraoxatridecan-1-amine | 85030-56-4 | MFCD11041099 | 98.8% | 207.27 | C9H21NO4 | 250 MG
SDP
Catalog No. 5000203879
Encompass_Preferred
m-PEG4-Amine is a polyethylene glycol (PEG) based bifunctional linker bearing a terminal primary amine. It is used as a short, flexible spacer in the synthesis of PROTACs and as a cleavable linker for antibody-drug conjugates, facilitating amine-based conjugation reactions.
  • Provides a PEG4 spacer (~4 ethylene glycol units)
  • Terminal primary amine for amide or carbamate conjugation
  • High purity (98.8%) suitable for research use
  • Molecular weight 207.27 g/mol
  • Typically supplied as a solid; common pack size 250 MG
Catalog No. 50-002-03879 Supplier Medchemexpress LLC Supplier No. HYW040214250MG
m-PEG4-Amine is a polyethylene glycol (PEG) based bifunctional linker bearing a terminal primary amine. It is used as a short, flexible spacer in the synthesis of PROTACs and as a cleavable linker for antibody-drug conjugates, facilitating amine-based conjugation reactions.
  • Provides a PEG4 spacer (~4 ethylene glycol units)
  • Terminal primary amine for amide or carbamate conjugation
  • High purity (98.8%) suitable for research use
  • Molecular weight 207.27 g/mol
  • Typically supplied as a solid; common pack size 250 MG