Learn More
Medchemexpress LLC 3,6,9,12-tetraoxatridecan-1-amine | 85030-56-4 | MFCD11041099 | 98.8% | 207.27 g/mol | C9H21NO4 | 500 MG
SDP
Catalog No. 5000203356
Encompass_Preferred
m-PEG4-Amine is a short polyethylene glycol (PEG4) amine reagent used as a linker in chemical biology applications. It is commonly used in the synthesis of proteolysis targeting chimeras (PROTACs) and can serve as a cleavable linker in antibody-drug conjugate (ADC) construction. The reagent is supplied at research-grade purity for laboratory use.
  • Short PEG4 amine linker for conjugation chemistry.
  • Useful for PROTAC design and linker optimization.
  • Functions as a cleavable linker for ADC synthesis.
  • High purity suitable for research applications.
  • Molecular weight 207.27 g/mol; formula C9H21NO4.
Catalog No. 50-002-03356 Supplier Medchemexpress LLC Supplier No. HYW040214500MG
m-PEG4-Amine is a short polyethylene glycol (PEG4) amine reagent used as a linker in chemical biology applications. It is commonly used in the synthesis of proteolysis targeting chimeras (PROTACs) and can serve as a cleavable linker in antibody-drug conjugate (ADC) construction. The reagent is supplied at research-grade purity for laboratory use.
  • Short PEG4 amine linker for conjugation chemistry.
  • Useful for PROTAC design and linker optimization.
  • Functions as a cleavable linker for ADC synthesis.
  • High purity suitable for research applications.
  • Molecular weight 207.27 g/mol; formula C9H21NO4.