Learn More
Medchemexpress LLC 3,6,9,12-tetraoxatridecan-1-ol | 23783-42-8 | 99.6% | 208.25 g/mol | C9H20O5 | 100 G
SDP
Catalog No. 5000300640
Encompass_Preferred
Tetraethylene glycol monomethyl ether is a low-molecular-weight polyethylene glycol derivative with a terminal methyl ether and a terminal hydroxyl. It functions as a flexible, hydrophilic spacer for linker design in PROTACs and other conjugation chemistries, helping to improve solubility and minimize steric hindrance. Typical properties: C9H20O5, 208.25 g/mol.
  • Provides flexible, hydrophilic spacer for linker design.
  • Improves solubility of conjugates in polar solvents.
  • Reduces steric hindrance between linked moieties.
  • Compatible with common synthetic transformations.
  • Available in multiple laboratory-scale package sizes.
Catalog No. 50-003-00640 Supplier Medchemexpress LLC Supplier No. HYW018365100G
Tetraethylene glycol monomethyl ether is a low-molecular-weight polyethylene glycol derivative with a terminal methyl ether and a terminal hydroxyl. It functions as a flexible, hydrophilic spacer for linker design in PROTACs and other conjugation chemistries, helping to improve solubility and minimize steric hindrance. Typical properties: C9H20O5, 208.25 g/mol.
  • Provides flexible, hydrophilic spacer for linker design.
  • Improves solubility of conjugates in polar solvents.
  • Reduces steric hindrance between linked moieties.
  • Compatible with common synthetic transformations.
  • Available in multiple laboratory-scale package sizes.