Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3,6,9,16-Tetraoxaheptadecanoic acid, 17-phenyl-, 1,1-dimethylethyl ester | 2381196-81-0 | 95.8% | 25 MG
SDP

Catalog No. 5000341443
Encompass_Preferred

Boc-C1-PEG3-C4-OBn (PROTAC Linker 15) is a PROTAC linker that refers to its PEG composition. This linker facilitates the synthesis of various PROTACs, which are molecules designed with two distinct ligands connected by the linker. One ligand targets an E3 ubiquitin ligase, while the other targets a specific protein of interest. PROTACs harness the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.

  • PROTAC linker with PEG composition
  • Facilitates the synthesis of various PROTACs
  • Designed to connect two distinct ligands for targeted protein degradation
  • Utilizes the ubiquitin-proteasome system for selective protein degradation
  • Molecular weight of 410.54
  • Liquid appearance, light yellow to yellow in color
  • Soluble in DMSO (100 mg/mL)

Catalog No. 50-003-41443 Supplier Medchemexpress LLC Supplier No. 25MGHY130619
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-C1-PEG3-C4-OBn (PROTAC Linker 15) is a PROTAC linker that refers to its PEG composition. This linker facilitates the synthesis of various PROTACs, which are molecules designed with two distinct ligands connected by the linker. One ligand targets an E3 ubiquitin ligase, while the other targets a specific protein of interest. PROTACs harness the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.

  • PROTAC linker with PEG composition
  • Facilitates the synthesis of various PROTACs
  • Designed to connect two distinct ligands for targeted protein degradation
  • Utilizes the ubiquitin-proteasome system for selective protein degradation
  • Molecular weight of 410.54
  • Liquid appearance, light yellow to yellow in color
  • Soluble in DMSO (100 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.