Learn More
Medchemexpress LLC 3,6,9,16-Tetraoxaheptadecanoic acid, 17-phenyl-, 1,1-dimethylethyl ester | 2381196-81-0 | 95.8% | 250 MG
SDP
Catalog No. 5000341506
Encompass_Preferred
Boc-C1-PEG3-C4-OBn (PROTAC Linker 15) is a PROTAC linker, characterized by its PEG composition. It is utilized in the synthesis of various PROTACs, which connect two distinct ligands with a linker. This mechanism leverages the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • PROTAC linker with PEG composition
  • Used in the synthesis of various PROTACs
  • Connects ligands for E3 ubiquitin ligase and target protein
  • Exploits intracellular ubiquitin-proteasome system
  • For research use only
Catalog No. 50-003-41506 Supplier Medchemexpress LLC Supplier No. 250MGHY130619
Boc-C1-PEG3-C4-OBn (PROTAC Linker 15) is a PROTAC linker, characterized by its PEG composition. It is utilized in the synthesis of various PROTACs, which connect two distinct ligands with a linker. This mechanism leverages the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • PROTAC linker with PEG composition
  • Used in the synthesis of various PROTACs
  • Connects ligands for E3 ubiquitin ligase and target protein
  • Exploits intracellular ubiquitin-proteasome system
  • For research use only