Learn More
Medchemexpress LLC 3,6,9-trioxa-12-azaheptadecan-17-oic acid, 1-amino-16-[[4-[[(2-amino-1,4-dihydro-4-oxo-6-pteridinyl)m | 710323-40-3 | 99.7% | 615.64 | C27H37N9O8 | 1 MG
SDP
Catalog No. 5000203719
Encompass_Preferred
Folate-PEG3-amine is a cleavable three-unit polyethylene glycol (PEG3) linker bearing a folate targeting moiety and a terminal amine for bioconjugation. It is intended for research use in the synthesis of antibody-drug conjugates and other targeted conjugates.
  • Cleavable three-unit PEG spacer enabling flexible linker length.
  • Terminal primary amine for straightforward conjugation chemistry.
  • Folate moiety for receptor-targeted delivery.
  • High purity (99.69%) suitable for analytical and synthetic workflows.
  • Molecular weight 615.64 and formula C27H37N9O8.
Catalog No. 50-002-03719 Supplier Medchemexpress LLC Supplier No. HY1384841MG
Folate-PEG3-amine is a cleavable three-unit polyethylene glycol (PEG3) linker bearing a folate targeting moiety and a terminal amine for bioconjugation. It is intended for research use in the synthesis of antibody-drug conjugates and other targeted conjugates.
  • Cleavable three-unit PEG spacer enabling flexible linker length.
  • Terminal primary amine for straightforward conjugation chemistry.
  • Folate moiety for receptor-targeted delivery.
  • High purity (99.69%) suitable for analytical and synthetic workflows.
  • Molecular weight 615.64 and formula C27H37N9O8.