Learn More
Medchemexpress LLC 3,6,9-trioxa-12-azaheptadecan-17-oic acid, folate conjugate | 710323-40-3 | 99.7% | 615.64 | C27H37N9O8 | 10 MG
SDP
Catalog No. 5000203803
Encompass_Preferred
Folate-PEG3-amine is a cleavable three-unit polyethylene glycol linker bearing a folate moiety, used for targeted conjugation and antibody-drug conjugate synthesis. It provides a hydrophilic spacer with an amine for coupling to drugs or biomolecules.
  • cleavable three-unit PEG spacer
  • folate targeting motif for receptor-mediated delivery
  • primary amine functional group for easy coupling
  • high purity suitable for research applications
  • hydrophilic spacer to improve solubility and reduce aggregation
Catalog No. 50-002-03803 Supplier Medchemexpress LLC Supplier No. HY13848410MG
Folate-PEG3-amine is a cleavable three-unit polyethylene glycol linker bearing a folate moiety, used for targeted conjugation and antibody-drug conjugate synthesis. It provides a hydrophilic spacer with an amine for coupling to drugs or biomolecules.
  • cleavable three-unit PEG spacer
  • folate targeting motif for receptor-mediated delivery
  • primary amine functional group for easy coupling
  • high purity suitable for research applications
  • hydrophilic spacer to improve solubility and reduce aggregation