Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3,6,9-Trioxaundecanedioic Acid

Catalog No. 92893250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
92-893-250ML 50 mL
92-893-50ML 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 92-893-250ML Supplier Sigma Aldrich Supplier No. 92893250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3,6,9-Trioxaundecanedioic Acid

Specifications

CAS 13887-98-4
Density 1.3 g/mL at 20°C
Molecular Formula C8H14O7
Refractive Index n20/D 1.470
Linear Formula C8H14O7
Quantity 50 mL
MDL Number MFCD00044099
Synonym O,O'-Oxydiethylene-diglycolic acid
Molecular Weight (g/mol) 222.19 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.