Learn More
Medchemexpress LLC 3,6-Acridinediamine, N3,N3,N6,N6-tetramethyl- | 494-38-2 | 96.58% | 500 MG
SDP
Catalog No. 5000410583
Encompass_Preferred
Acridine Orange base is a cell-permeable fluorescent dye. It stains various organisms such as bacteria, parasites, and viruses bright orange. When used under appropriate conditions (pH=3.5, Ex=460 nm), it can distinguish human cells in green for detection by fluorescence microscopy. This dye fluoresces green when bound to dsDNA (Ex=488, Em=520-524) and red when bound to ssDNA (Ex=457, Em=630-644) or ssRNA (Ex=457, Em=630-644). It can also be utilized in cell cycle assays.
  • Cell-permeable fluorescent dye.
  • Stains organisms bright orange.
  • Distinguishes human cells in green under specific conditions.
  • Fluoresces green when bound to dsDNA.
  • Fluoresces red when bound to ssDNA or ssRNA.
  • Can be used in cell cycle assays.
Catalog No. 50-004-10583 Supplier Medchemexpress LLC Supplier No. HYD0952500MG
Acridine Orange base is a cell-permeable fluorescent dye. It stains various organisms such as bacteria, parasites, and viruses bright orange. When used under appropriate conditions (pH=3.5, Ex=460 nm), it can distinguish human cells in green for detection by fluorescence microscopy. This dye fluoresces green when bound to dsDNA (Ex=488, Em=520-524) and red when bound to ssDNA (Ex=457, Em=630-644) or ssRNA (Ex=457, Em=630-644). It can also be utilized in cell cycle assays.
  • Cell-permeable fluorescent dye.
  • Stains organisms bright orange.
  • Distinguishes human cells in green under specific conditions.
  • Fluoresces green when bound to dsDNA.
  • Fluoresces red when bound to ssDNA or ssRNA.
  • Can be used in cell cycle assays.