Learn More
Medchemexpress LLC 3-(6-bromo-1-oxoisoindolin-2-yl)piperidine-2,6-dione | 2304513-76-4 | MFCD32667217 | 99.9% | 323.15 g/mol | C13H11BrN2O3 | 25 G
SDP
Catalog No. 5000280819
Encompass_Preferred
3-(6-Bromo-1-oxoisoindolin-2-yl)piperidine-2,6-dione is a lenalidomide analog used as a building block in targeted protein degradation (PROTAC) research. The compound contains a brominated isoindolinone core fused to a 2,6-dioxopiperidine moiety, is supplied as a solid, and is provided at high stated purity for chemical biology applications.
  • Lenalidomide analog for PROTAC research.
  • Brominated isoindolinone core for ligand design.
  • High purity (99.85%) suitable for analytical use.
  • Molecular formula C13H11BrN2O3; molecular weight 323.15 g/mol.
  • Solid, gray to gray purple appearance.
  • Stable at room temperature for up to 3 years; follow solvent storage recommendations.
Catalog No. 50-002-80819 Supplier Medchemexpress LLC Supplier No. HYW24340425G
3-(6-Bromo-1-oxoisoindolin-2-yl)piperidine-2,6-dione is a lenalidomide analog used as a building block in targeted protein degradation (PROTAC) research. The compound contains a brominated isoindolinone core fused to a 2,6-dioxopiperidine moiety, is supplied as a solid, and is provided at high stated purity for chemical biology applications.
  • Lenalidomide analog for PROTAC research.
  • Brominated isoindolinone core for ligand design.
  • High purity (99.85%) suitable for analytical use.
  • Molecular formula C13H11BrN2O3; molecular weight 323.15 g/mol.
  • Solid, gray to gray purple appearance.
  • Stable at room temperature for up to 3 years; follow solvent storage recommendations.