Learn More
Medchemexpress LLC 3-(6-chloropyridine-3-yloxymethyl)-2-azabicyclo[3.1.0]hexane hydrochloride | 2414674-71-6 | 99.4% | 261.15 | C12H12ClN2O | 5 MG
SDP
Catalog No. 5000222100
Encompass_Preferred
SUVN-911 is a potent, selective, brain-penetrant, orally bioavailable neuronal nicotinic acetylcholine α4β2 receptor antagonist (hydrochloride salt) with reported antidepressant activity. Supplied as a high-purity solid for research use in in vitro and in vivo studies.
  • High potency at α4β2 receptors (Ki 1.5 nM).
  • High selectivity over other nAChR subtypes and common off-targets.
  • Brain-penetrant and orally bioavailable, enabling animal pharmacology studies.
  • High-purity solid form for reproducible experimental results.
  • Soluble in DMSO (~125 mg/mL); powder storage at 4°C, solutions recommended at -80°C.
Catalog No. 50-002-22100 Supplier Medchemexpress LLC Supplier No. HY1361465MG
SUVN-911 is a potent, selective, brain-penetrant, orally bioavailable neuronal nicotinic acetylcholine α4β2 receptor antagonist (hydrochloride salt) with reported antidepressant activity. Supplied as a high-purity solid for research use in in vitro and in vivo studies.
  • High potency at α4β2 receptors (Ki 1.5 nM).
  • High selectivity over other nAChR subtypes and common off-targets.
  • Brain-penetrant and orally bioavailable, enabling animal pharmacology studies.
  • High-purity solid form for reproducible experimental results.
  • Soluble in DMSO (~125 mg/mL); powder storage at 4°C, solutions recommended at -80°C.