Learn More
Medchemexpress LLC 3-(6-chloropyridine-3-yloxymethyl)-2-azabicyclo[3.1.0]hexane hydrochloride | 2414674-71-6 | 99.4% | C11H14Cl2N2O | 10MG
SDP
Catalog No. 5000260185
Encompass_Preferred
This small-molecule hydrochloride is a potent, selective, brain-penetrant, and orally bioavailable neuronal nicotinic acetylcholine α4β2 receptor antagonist with a reported Ki ≈ 1.5 nM and demonstrated antidepressant activity in preclinical models. It is described in the literature and supplier listings as a crystalline, non-hygroscopic salt with favorable ADME and safety properties.
  • Potent, selective α4β2 receptor antagonism (Ki ≈ 1.5 nM).
  • Oral bioavailability and brain penetration demonstrated in preclinical studies.
  • Favorable ADME profile with low drug-drug interaction liability.
  • Crystalline hydrochloride salt with stable physicochemical properties.
  • Reported high purity in supplier analyses (example: 99.4%).
  • Supported by published preclinical studies and supplier data.
Catalog No. 50-002-60185 Supplier Medchemexpress LLC Supplier No. HY13614610MG
This small-molecule hydrochloride is a potent, selective, brain-penetrant, and orally bioavailable neuronal nicotinic acetylcholine α4β2 receptor antagonist with a reported Ki ≈ 1.5 nM and demonstrated antidepressant activity in preclinical models. It is described in the literature and supplier listings as a crystalline, non-hygroscopic salt with favorable ADME and safety properties.
  • Potent, selective α4β2 receptor antagonism (Ki ≈ 1.5 nM).
  • Oral bioavailability and brain penetration demonstrated in preclinical studies.
  • Favorable ADME profile with low drug-drug interaction liability.
  • Crystalline hydrochloride salt with stable physicochemical properties.
  • Reported high purity in supplier analyses (example: 99.4%).
  • Supported by published preclinical studies and supplier data.