Learn More
Medchemexpress LLC 3,6-Di-tert-butyl-9H-carbazole-d25 | 2569012-64-0 | 304.57 | 50 MG
SDP
Catalog No. 5000455034
Encompass_Preferred
3,6-Di-tert-butyl-9H-carbazole-d25 is a deuterium labeled 3,6-Di-tert-butyl-9H-carbazole. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Potential to affect the pharmacokinetic and metabolic profiles of drugs due to deuteration.
Catalog No. 50-004-55034 Supplier Medchemexpress LLC Supplier No. HYW011565S150MG
3,6-Di-tert-butyl-9H-carbazole-d25 is a deuterium labeled 3,6-Di-tert-butyl-9H-carbazole. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Potential to affect the pharmacokinetic and metabolic profiles of drugs due to deuteration.