Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,6-Diaminocarbazole 98.0+%, TCI America™
SDP

Catalog No. D21165G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D21165G 5 g
D21161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D21165G Supplier TCI America Supplier No. D21165G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 86-71-5
Molecular Formula C12H11N3
Molecular Weight (g/mol) 197.24
MDL Number MFCD00143081
InChI Key YCZUWQOJQGCZKG-UHFFFAOYSA-N
Synonym 9H-Carbazole-3,6-diamine
PubChem CID 47108
IUPAC Name 9H-carbazole-3,6-diamine
SMILES NC1=CC2=C(NC3=C2C=C(N)C=C3)C=C1

Specifications

Quantity 5 g
UN Number 3259
Formula Weight 197.24
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Diaminocarbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.