Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3,6-diazabicyclo[3.2.2]nonane, 3-[(4-methyl-1h-imidazol-5-yl)methyl] (dihydrochloride) | 1453028-33-5 | 98.6% | 439.38 g/mol | C20H28Cl2N6O | 10 MG
SDP

Catalog No. 5000286113
Encompass_Preferred

DRP1i27 dihydrochloride is a research-grade small-molecule inhibitor of dynamin-related protein 1 (Drp1), provided as the dihydrochloride salt for laboratory studies of mitochondrial fission and related cellular processes.

  • Potent inhibitor of Drp1 GTPase activity demonstrated in cell models.
  • Supplied as a dihydrochloride salt suitable for biochemical and cellular assays.
  • Available in multiple quantities and formats, including solid and 10 mM DMSO solutions.
  • High purity: 98.6% (reported by manufacturer).
  • Molecular formula C20H28Cl2N6O; molecular weight 439.38 g/mol.
  • Documentation available: datasheet, COA, SDS, and spectral data for QC support.

Catalog No. 50-002-86113 Supplier Medchemexpress LLC Supplier No. HY152086A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DRP1i27 dihydrochloride is a research-grade small-molecule inhibitor of dynamin-related protein 1 (Drp1), provided as the dihydrochloride salt for laboratory studies of mitochondrial fission and related cellular processes.

  • Potent inhibitor of Drp1 GTPase activity demonstrated in cell models.
  • Supplied as a dihydrochloride salt suitable for biochemical and cellular assays.
  • Available in multiple quantities and formats, including solid and 10 mM DMSO solutions.
  • High purity: 98.6% (reported by manufacturer).
  • Molecular formula C20H28Cl2N6O; molecular weight 439.38 g/mol.
  • Documentation available: datasheet, COA, SDS, and spectral data for QC support.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.