Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,6-Dibromo-9-(4-dodecyloxyphenyl)-9H-carbazole 97.0+%, TCI America™
SDP

Catalog No. D5343200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5343200MG 200 mg
D53431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5343200MG Supplier TCI America Supplier No. D5343200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 865163-47-9
Molecular Formula C30H35Br2NO
Molecular Weight (g/mol) 585.424
InChI Key VWCTWVHXOOAKSE-UHFFFAOYSA-N
PubChem CID 86071764
IUPAC Name 3,6-dibromo-9-(4-dodecoxyphenyl)carbazole
SMILES CCCCCCCCCCCCOC1=CC=C(C=C1)N2C3=C(C=C(C=C3)Br)C4=C2C=CC(=C4)Br

Specifications

Melting Point 91°C
Color Yellow
Quantity 200 mg
Formula Weight 585.42
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Dibromo-9-(4-dodecyloxyphenyl)-9H-carbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.