Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,6-Dibromo-9H-fluoren-9-one 95.0+%, TCI America™
SDP

Catalog No. D53011G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D53011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D53011G Supplier TCI America Supplier No. D53011G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 216312-73-1
Molecular Formula C13H6Br2O
Molecular Weight (g/mol) 338.00
MDL Number MFCD30478800
InChI Key XXZOHYHKWDLSFS-UHFFFAOYSA-N
PubChem CID 257937
IUPAC Name 3,6-dibromo-9H-fluoren-9-one
SMILES BrC1=CC2=C(C=C1)C(=O)C1=C2C=C(Br)C=C1

Specifications

Melting Point 324°C
Quantity 1 g
Formula Weight 338.00
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Dibromo-9H-fluoren-9-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.