Learn More
Medchemexpress LLC 3,6-Dibromocarbazole | 6825-20-3 | 99.97% | 325.00 | 25 G
SDP
Catalog No. 5000366619
Encompass_Preferred
3,6-Dibromocarbazole is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is suitable for a variety of laboratory applications.
  • Biochemical reagent
  • Organic compound for life science related research
  • For research use only
  • Shipping at room temperature in continental US
  • Store powder at -20°C for 3 years, 4°C for 2 years
  • Store in solvent at -80°C for 6 months, -20°C for 1 month
  • Soluble in DMSO at 200 mg/mL (615.38 mM)
Catalog No. 50-003-66619 Supplier Medchemexpress LLC Supplier No. 25GHYW009154
3,6-Dibromocarbazole is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is suitable for a variety of laboratory applications.
  • Biochemical reagent
  • Organic compound for life science related research
  • For research use only
  • Shipping at room temperature in continental US
  • Store powder at -20°C for 3 years, 4°C for 2 years
  • Store in solvent at -80°C for 6 months, -20°C for 1 month
  • Soluble in DMSO at 200 mg/mL (615.38 mM)