Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,6-Dibromothieno[3,2-b]thiophene 98.0+%, TCI America™
SDP

Catalog No. D36791G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36791G 1 g
D3679200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D36791G Supplier TCI America Supplier No. D36791G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 392662-65-6
Molecular Formula C6H2Br2S2
Molecular Weight (g/mol) 298.01
MDL Number MFCD10000958
InChI Key DFMPVYZMICAYCS-UHFFFAOYSA-N
PubChem CID 10613979
IUPAC Name 3,6-dibromothieno[3,2-b]thiophene
SMILES BrC1=CSC2=C1SC=C2Br

Specifications

Melting Point 127°C
Color White-Yellow
Quantity 1 g
Formula Weight 298.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Dibromothieno[3,2-b]thiophene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.