Learn More
Medchemexpress LLC 3,6-dichloropyridine-2-carboxylic acid | 1702-17-6 | MFCD00078655 | 98.5% | 192.00 g/mol | C6H3Cl2NO2 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW018749
Encompass_Preferred
Clopyralid (3,6-dichloropyridine-2-carboxylic acid) is a selective pyridine herbicide supplied as a research-grade reagent for analytical, formulation, and experimental use. Supplied as a high-purity solid (CAS 1702-17-6), it is used in studies of herbicidal activity, environmental fate, and formulation development; consult the certificate of analysis and safety data sheet for handling and storage.
  • High purity (98.52%), suitable for analytical applications.
  • Confirmed identity as 3,6-dichloropyridine-2-carboxylic acid (CAS 1702-17-6).
  • Available in multiple pack sizes, including 25 g.
  • Appropriate for herbicide activity, toxicology, and environmental fate studies.
  • Certificate of analysis and safety data sheet available for batch-specific information.
Catalog No. 50-003-04163