Learn More
AdipoGen 3 6-Dichlorotrimellitic acid
SDP
Catalog No. 502561158
Encompass_Preferred
Chemical. CAS 137071-78-4. Formula C9H4Cl2O6. MW 279.03. Synthetic. Key precursor used for preparation of a variety of dichlorinated fluoresceins and rhodamines such as TET and HEX. These chlorinated fluoresceins and rhodamines are widely used for labeling oligos and in DNA sequencing.
Catalog No. 50-256-1158 Supplier AdipoGen Supplier No. CDXD0268G001
Chemical. CAS 137071-78-4. Formula C9H4Cl2O6. MW 279.03. Synthetic. Key precursor used for preparation of a variety of dichlorinated fluoresceins and rhodamines such as TET and HEX. These chlorinated fluoresceins and rhodamines are widely used for labeling oligos and in DNA sequencing.