Learn More
Medchemexpress LLC 3',6'-dihydroxy-3-oxo-N-prop-2-ynylspiro[2-benzofuran-1,9'-xanthene]-5-carboxamide | 510758-19-7 | MFCD28505571 | 95.0% | C24H15NO6 | 10MG
SDP
Catalog No. 5000261889
Encompass_Preferred
5-FAM-Alkyne is an alkyne-functionalized fluorescein derivative supplied as a solid fluorescent labeling reagent for copper-catalyzed click chemistry. It is designed for conjugation to biomolecules and use in fluorescence-based detection and imaging applications, providing reliable labeling in typical biochemical workflows.
  • Alkyne functional group for click chemistry.
  • High purity (95.0%).
  • Solid form, yellow to brown.
  • Molecular weight 413.38 g/mol.
  • Suitable for fluorescent labeling and imaging applications.
Catalog No. 50-002-61889 Supplier Medchemexpress LLC Supplier No. HY13091310MG
5-FAM-Alkyne is an alkyne-functionalized fluorescein derivative supplied as a solid fluorescent labeling reagent for copper-catalyzed click chemistry. It is designed for conjugation to biomolecules and use in fluorescence-based detection and imaging applications, providing reliable labeling in typical biochemical workflows.
  • Alkyne functional group for click chemistry.
  • High purity (95.0%).
  • Solid form, yellow to brown.
  • Molecular weight 413.38 g/mol.
  • Suitable for fluorescent labeling and imaging applications.