Learn More
Medchemexpress LLC 3,6-Dihydroxyflavone | 108238-41-1 | 10 MG
SDP
Catalog No. 5000417277
Encompass_Preferred
3,6-Dihydroxyflavone is an anti-cancer agent that dose- and time-dependently decreases cell viability and induces apoptosis by activating the caspase cascade and cleaving poly (ADP-ribose) polymerase (PARP). It also increases intracellular oxidative stress and lipid peroxidation.
  • Anti-cancer agent
  • Decreases cell viability
  • Induces apoptosis by activating caspase cascade
  • Cleaves poly (ADP-ribose) polymerase (PARP)
  • Increases intracellular oxidative stress
  • Increases lipid peroxidation
Catalog No. 50-004-17277 Supplier Medchemexpress LLC Supplier No. HYN848110MG
3,6-Dihydroxyflavone is an anti-cancer agent that dose- and time-dependently decreases cell viability and induces apoptosis by activating the caspase cascade and cleaving poly (ADP-ribose) polymerase (PARP). It also increases intracellular oxidative stress and lipid peroxidation.
  • Anti-cancer agent
  • Decreases cell viability
  • Induces apoptosis by activating caspase cascade
  • Cleaves poly (ADP-ribose) polymerase (PARP)
  • Increases intracellular oxidative stress
  • Increases lipid peroxidation