Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3,6-Dihydroxyflavone | 108238-41-1 | 25 MG
SDP

Catalog No. 5000421475
Encompass_Preferred

3,6-Dihydroxyflavone is an anti-cancer agent that decreases cell viability and induces apoptosis. It works by activating the caspase cascade and cleaving poly (ADP-ribose) polymerase (PARP). Additionally, it increases intracellular oxidative stress and lipid peroxidation.

  • Decreases cell viability and induces apoptosis
  • Activates caspase cascade
  • Cleaves poly (ADP-ribose) polymerase (PARP)
  • Increases intracellular oxidative stress
  • Increases lipid peroxidation
  • Appears as a light yellow to light brown solid
  • Has a molecular weight of 254.24
  • Chemical formula is C15H10O4
  • Purity is 98.06%
  • Soluble in DMSO at 66.67 mg/mL

Catalog No. 50-004-21475 Supplier Medchemexpress LLC Supplier No. 25MGHYN8481
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3,6-Dihydroxyflavone is an anti-cancer agent that decreases cell viability and induces apoptosis. It works by activating the caspase cascade and cleaving poly (ADP-ribose) polymerase (PARP). Additionally, it increases intracellular oxidative stress and lipid peroxidation.

  • Decreases cell viability and induces apoptosis
  • Activates caspase cascade
  • Cleaves poly (ADP-ribose) polymerase (PARP)
  • Increases intracellular oxidative stress
  • Increases lipid peroxidation
  • Appears as a light yellow to light brown solid
  • Has a molecular weight of 254.24
  • Chemical formula is C15H10O4
  • Purity is 98.06%
  • Soluble in DMSO at 66.67 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.