Learn More
Medchemexpress LLC 3,6-dihydroxyxanthone | 1214-24-0 | MFCD00226967 | 99.6% | 228.20 g·mol⁻¹ | C13H8O4 | 250 MG
SDP
Catalog No. 5000303427
Encompass_Preferred
3,6-Dihydroxyxanthone is a xanthone derivative provided for research use and reported to exhibit anticancer and antioxidant activities. It is intended for biochemical and cell biology studies investigating cytotoxicity, enzyme inhibition, and related cancer research applications.
  • High chemical purity suitable for research (99.6%).
  • Reported anticancer and antioxidant activity useful for mechanism studies.
  • Molecular formula C13H8O4; molecular weight 228.20 g·mol⁻¹.
  • Available in multiple quantities to support small-scale and preparative work.
  • Solid form recommended storage at 4°C under inert atmosphere; solutions require low-temperature storage.
  • Suitable for preparation of DMSO solutions for biological assays.
Catalog No. 50-003-03427 Supplier Medchemexpress LLC Supplier No. 250MGHYW031510
3,6-Dihydroxyxanthone is a xanthone derivative provided for research use and reported to exhibit anticancer and antioxidant activities. It is intended for biochemical and cell biology studies investigating cytotoxicity, enzyme inhibition, and related cancer research applications.
  • High chemical purity suitable for research (99.6%).
  • Reported anticancer and antioxidant activity useful for mechanism studies.
  • Molecular formula C13H8O4; molecular weight 228.20 g·mol⁻¹.
  • Available in multiple quantities to support small-scale and preparative work.
  • Solid form recommended storage at 4°C under inert atmosphere; solutions require low-temperature storage.
  • Suitable for preparation of DMSO solutions for biological assays.