Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,6-Diiodo-9-phenylcarbazole 98.0+%, TCI America™
SDP

Catalog No. D45431G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45431G 1 g
D45435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D45431G Supplier TCI America Supplier No. D45431G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 57103-21-6
Molecular Formula C18H11I2N
Molecular Weight (g/mol) 495.102
MDL Number MFCD11521285
InChI Key AWGAUYXFWGUFNE-UHFFFAOYSA-N
PubChem CID 13407347
IUPAC Name 3,6-diiodo-9-phenylcarbazole
SMILES C1=CC=C(C=C1)N2C3=C(C=C(C=C3)I)C4=C2C=CC(=C4)I

Specifications

Melting Point 183°C
Quantity 1 g
Formula Weight 495.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Diiodo-9-phenylcarbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.