Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3,6-diiodo-9H-carbazole | 57103-02-3 | MFCD00450017 | 99.6% | 419.00 | C12H7I2N | 10 G
SDP

Catalog No. 5000301240
Encompass_Preferred

3,6-Diiodo-9H-carbazole is an iodinated carbazole derivative supplied as a solid reagent for research use. It functions as a versatile building block and intermediate in organic synthesis and medicinal chemistry for constructing iodinated carbazole scaffolds and derivatives.

  • High purity (99.6%).
  • Molecular formula C12H7I2N.
  • Molecular weight 419.00 g·mol⁻¹.
  • Supplied as a solid powder for easy handling and storage.
  • Suitable for organic synthesis and medicinal chemistry research.
  • Available in multiple pack sizes for scale flexibility.
  • Comes with supporting documentation such as SDS and COA.

Catalog No. 50-003-01240 Supplier Medchemexpress LLC Supplier No. HYW02222410G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3,6-Diiodo-9H-carbazole is an iodinated carbazole derivative supplied as a solid reagent for research use. It functions as a versatile building block and intermediate in organic synthesis and medicinal chemistry for constructing iodinated carbazole scaffolds and derivatives.

  • High purity (99.6%).
  • Molecular formula C12H7I2N.
  • Molecular weight 419.00 g·mol⁻¹.
  • Supplied as a solid powder for easy handling and storage.
  • Suitable for organic synthesis and medicinal chemistry research.
  • Available in multiple pack sizes for scale flexibility.
  • Comes with supporting documentation such as SDS and COA.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.