Learn More
Medchemexpress LLC 3,6-diiodo-9H-carbazole | 57103-02-3 | MFCD00450017 | 99.6% | 419.00 g/mol | C12H7I2N | 5 G
SDP
Catalog No. 5000299600
Encompass_Preferred
3,6-Diiodo-9H-carbazole is an iodinated heterocyclic compound used as a building block in organic synthesis and medicinal chemistry research. It is commonly employed to prepare carbazole derivatives and to introduce iodine-functionalized positions for subsequent cross-coupling or substitution reactions.
  • High purity suitable for research and synthesis.
  • Iodinated positions facilitate cross-coupling and substitution reactions.
  • Solid form enables stable storage and handling.
  • Available in multiple pack sizes to support scale flexibility.
  • Characterized by formula C12H7I2N and molecular weight 419.00 g/mol.
Catalog No. 50-002-99600 Supplier Medchemexpress LLC Supplier No. HYW0222245G
3,6-Diiodo-9H-carbazole is an iodinated heterocyclic compound used as a building block in organic synthesis and medicinal chemistry research. It is commonly employed to prepare carbazole derivatives and to introduce iodine-functionalized positions for subsequent cross-coupling or substitution reactions.
  • High purity suitable for research and synthesis.
  • Iodinated positions facilitate cross-coupling and substitution reactions.
  • Solid form enables stable storage and handling.
  • Available in multiple pack sizes to support scale flexibility.
  • Characterized by formula C12H7I2N and molecular weight 419.00 g/mol.