Learn More
Medchemexpress LLC 3,6-diiodo-9H-carbazole | 57103-02-3 | MFCD00450017 | 99.6% | 419.00 g/mol | C12H7I2N | 50 G
SDP
Catalog No. 5000301591
Encompass_Preferred
3,6-diiodo-9H-carbazole is an iodinated carbazole derivative supplied for research use as a synthetic building block in organic and medicinal chemistry. It has molecular formula C12H7I2N and a molecular weight of 419.00 g/mol. The compound is provided as a solid with high purity and is intended for use in the design and synthesis of carbazole derivatives; store at 4°C protected from light.
  • High purity (99.64%) suitable for research applications.
  • Solid physical form for solid-state handling.
  • Useful building block in organic and medicinal chemistry synthesis.
  • Stable under recommended storage conditions (4°C, protect from light).
  • Available in multiple package sizes for scale flexibility.
Catalog No. 50-003-01591 Supplier Medchemexpress LLC Supplier No. HYW02222450G
3,6-diiodo-9H-carbazole is an iodinated carbazole derivative supplied for research use as a synthetic building block in organic and medicinal chemistry. It has molecular formula C12H7I2N and a molecular weight of 419.00 g/mol. The compound is provided as a solid with high purity and is intended for use in the design and synthesis of carbazole derivatives; store at 4°C protected from light.
  • High purity (99.64%) suitable for research applications.
  • Solid physical form for solid-state handling.
  • Useful building block in organic and medicinal chemistry synthesis.
  • Stable under recommended storage conditions (4°C, protect from light).
  • Available in multiple package sizes for scale flexibility.