Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,6-Diiodocarbazole 98.0+%, TCI America™
SDP

Catalog No. D45485G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45485G 5 g
D454825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D45485G Supplier TCI America Supplier No. D45485G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 57103-02-3
Molecular Formula C12H7I2N
Molecular Weight (g/mol) 419.00
MDL Number MFCD00450017
InChI Key PECAOKZHORDWAI-UHFFFAOYSA-N
PubChem CID 3116488
IUPAC Name 3,6-diiodo-9H-carbazole
SMILES IC1=CC2=C(NC3=C2C=C(I)C=C3)C=C1

Specifications

Melting Point 207°C
Quantity 5 g
Formula Weight 419.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Diiodocarbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.