Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,6-Dimethyl-1-heptyn-3-ol 98.0+%, TCI America™
SDP

Catalog No. D12665ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D12665ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D12665ML Supplier TCI America Supplier No. D12665ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 19549-98-5
Molecular Formula C9H16O
Molecular Weight (g/mol) 140.23
MDL Number MFCD00041608
InChI Key COWFALGRNJAHKW-UHFFFAOYNA-N
Synonym (1,4-Dimethyl-1-hydroxypentyl)acetylene
PubChem CID 89182
IUPAC Name 3,6-dimethylhept-1-yn-3-ol
SMILES CC(C)CCC(C)(O)C#C

Specifications

Color Yellow
Boiling Point 76°C
Quantity 5 mL
UN Number 1987
Formula Weight 140.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,6-Dimethyl-1-heptyn-3-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.