Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3,6-Dimethyl-1-p-tolyl-1H-pyrazolo[3,4-b]pyridine-4-carboxylic acid

Catalog No. TMT005521G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
TMT005521G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. TMT005521G Supplier Sigma Aldrich Supplier No. TMT005521G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant for synthesis of: Milrinone analogs to analyze effects on intracellular calcium increase in cardiac cells Tetrahydronaphthalene derivatives Quorum sensing modulators Selective anti-Helicobacter pylori activity molecules Orally active M1 mAChR agonists Aza derivatives of phytoalexin for antibacterial activity

Specifications

Molecular Formula C16H15N3O2
Linear Formula C16H15N3O2
MDL Number MFCD12547979
Quantity 25 g
Molecular Weight (g/mol) 281.31
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.