Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3,6-Dimethyl-9H-carbazole

Catalog No. UPL00291G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
UPL00291G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. UPL00291G Supplier Sigma Aldrich Supplier No. UPL00291G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Supersonic jet fluorescence spectra of 1,3-benzodioxole has been studied
  • The far-infrared spectrum of the vapour of 1,3-benzodioxole has been reported
  • The electronic absorption spectra and the laser-induced fluorescence spectra of supersonic-jet-cooled 1,3-benzodioxole molecules has been investigated

Specifications

CAS 5599-50-8
Molecular Formula C14H13N
Linear Formula C14H13N
MDL Number MFCD00814049
Quantity 50 g
Molecular Weight (g/mol) 195.26
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.