Learn More
At Battery Company Inc At Battery Company Inc Saft LS17500 A 3.6V Primary Lithium Battery, 480/cs
SDP
Catalog No. NC3761656
Encompass_Preferred
Saft LS17500 primary lithium cell, is based on the Lithium-Thionyl chloride (Li-SOCl2) chemistry. The LS17500 key features are the concentric electrodes (€œbobbin€) construction that enables maximum loading of active materials and corresponding energy.The LS17500 cell is designed specifically for long-term (3 to 15 years) applications, featuring a few UA based currents and periodic pulses, typically in the 5-150 mA range.
Catalog No. NC3761656 Supplier At Battery Company Inc Supplier No. LS17500
Add to Cart
Add to Cart
Request a Quote
Saft LS17500 primary lithium cell, is based on the Lithium-Thionyl chloride (Li-SOCl2) chemistry. The LS17500 key features are the concentric electrodes (€œbobbin€) construction that enables maximum loading of active materials and corresponding energy.The LS17500 cell is designed specifically for long-term (3 to 15 years) applications, featuring a few UA based currents and periodic pulses, typically in the 5-150 mA range.