Learn More
Medchemexpress LLC 3,7,4'-Trihydroxyflavone | 2034-65-3 | 99.4% | 270.24 | 1 ML
SDP
Catalog No. 5000438943
Encompass_Preferred
3,7,4'-Trihydroxyflavone, also known as 5-Deoxykampferol, is a flavonoid compound isolated from the stems of Rhus javanica var. roxburghiana. It exhibits anti-inflammatory and antioxidant properties by scavenging various reactive oxygen species (ROS) and reactive nitrogen species (RNS), and can also cleave DNA in the presence of copper ions.
  • Isolated from natural sources like Rhus javanica var. roxburghiana.
  • Functions as an anti-inflammatory agent.
  • Possesses antioxidant activities.
  • Capable of scavenging reactive oxygen and nitrogen species.
  • Can cleave DNA when copper ions are present.
Catalog No. 50-004-38943 Supplier Medchemexpress LLC Supplier No. HY11180610MM/1ML
3,7,4'-Trihydroxyflavone, also known as 5-Deoxykampferol, is a flavonoid compound isolated from the stems of Rhus javanica var. roxburghiana. It exhibits anti-inflammatory and antioxidant properties by scavenging various reactive oxygen species (ROS) and reactive nitrogen species (RNS), and can also cleave DNA in the presence of copper ions.
  • Isolated from natural sources like Rhus javanica var. roxburghiana.
  • Functions as an anti-inflammatory agent.
  • Possesses antioxidant activities.
  • Capable of scavenging reactive oxygen and nitrogen species.
  • Can cleave DNA when copper ions are present.