Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3,7-bis(4-methoxybenzyl)-3,7-dihydro-2H-diimidazo[4,5-d:4',5'-f][1,3]diazepin-2-one | 1070773-09-9 | MFCD30489740 | 99.6% | 428.44 g·mol⁻¹ | C23H20N6O3 | 1 ML
SDP

Catalog No. 5000214742
Encompass_Preferred

RK-33 is a small-molecule inhibitor of the RNA helicase DDX3 used for preclinical and molecular biology research. It inhibits DDX3 helicase activity and is intended for laboratory studies investigating RNA processing, translation, cell-cycle regulation, and related pathways.

  • Inhibits DDX3 RNA helicase activity for mechanistic studies.
  • Available as a 10 mM solution in DMSO (1 mL) suitable for in vitro assays.
  • High purity (≈99.6%) to minimize off-target effects.
  • Soluble in DMSO and formulatable for in vivo dosing with PEG300/saline.
  • Recommended storage conditions for powder and solutions to preserve stability.

Catalog No. 50-002-14742 Supplier Medchemexpress LLC Supplier No. HY10045510MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RK-33 is a small-molecule inhibitor of the RNA helicase DDX3 used for preclinical and molecular biology research. It inhibits DDX3 helicase activity and is intended for laboratory studies investigating RNA processing, translation, cell-cycle regulation, and related pathways.

  • Inhibits DDX3 RNA helicase activity for mechanistic studies.
  • Available as a 10 mM solution in DMSO (1 mL) suitable for in vitro assays.
  • High purity (≈99.6%) to minimize off-target effects.
  • Soluble in DMSO and formulatable for in vivo dosing with PEG300/saline.
  • Recommended storage conditions for powder and solutions to preserve stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.