Learn More
Medchemexpress LLC 3-(7-bromo-1-oxoisoindolin-2-yl)piperidine-2,6-dione | 2229976-08-1 | 99.0% | 323.15 g/mol | C13H11BrN2O3 | 1 G
SDP
Catalog No. 5000282357
Encompass_Preferred
3-(7-bromo-1-oxoisoindolin-2-yl)piperidine-2,6-dione is a lenalidomide analog used as a building block and E3 ligase ligand in PROTAC research. The compound is reported with formula C13H11BrN2O3, molecular weight 323.15 g/mol, and CAS 2229976-08-1. Analytical documentation is provided to support research use.
Catalog No. 50-002-82357 Supplier Medchemexpress LLC Supplier No. HYW2434051G
Add to Cart
Add to Cart
3-(7-bromo-1-oxoisoindolin-2-yl)piperidine-2,6-dione is a lenalidomide analog used as a building block and E3 ligase ligand in PROTAC research. The compound is reported with formula C13H11BrN2O3, molecular weight 323.15 g/mol, and CAS 2229976-08-1. Analytical documentation is provided to support research use.