Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,7-Diamino-2,8-dimethyldibenzothiophene Sulfone (contains 2,6-Dimethyl isomer) 70.0+%, TCI America™
SDP

Catalog No. T141925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T141925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T141925G Supplier TCI America Supplier No. T141925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 55011-44-4
Molecular Formula C14H14N2O2S
Molecular Weight (g/mol) 274.338
MDL Number MFCD00039157
InChI Key OJSPYCPPVCMEBS-UHFFFAOYSA-N
Synonym 3,7-Diamino-2,8-dimethyldiphenylenesulfone, o-Tolidine Sulfone
PubChem CID 4616894
IUPAC Name 2,8-dimethyl-5,5-dioxodibenzothiophene-3,7-diamine
SMILES CC1=C(C=C2C(=C1)C3=CC(=C(C=C3S2(=O)=O)N)C)N

Specifications

Color Yellow
Quantity 25 g
Formula Weight 274.34
Percent Purity ≥70.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 3,7-Diamino-2,8-dimethyldibenzothiophene Sulfone (contains 2,6-Dimethyl isomer)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.