Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,7-Dibromodibenzo[b,d]thiophene 96.0+%, TCI America™
SDP

Catalog No. D4274200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4274200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4274200MG Supplier TCI America Supplier No. D4274200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 83834-10-0
Molecular Formula C12H6Br2S
Molecular Weight (g/mol) 342.048
MDL Number MFCD01027447
InChI Key KMNBVODPWIFUSS-UHFFFAOYSA-N
PubChem CID 803759
IUPAC Name 3,7-dibromodibenzothiophene
SMILES C1=CC2=C(C=C1Br)SC3=C2C=CC(=C3)Br

Specifications

Melting Point 177°C
Color White-Yellow
Quantity 200 mg
Formula Weight 342.05
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,7-Dibromodibenzo[b,d]thiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.