Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,7-Dithia-1,9-nonanedithiol 97.0+%, TCI America™
SDP

Catalog No. D28521G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28521G 1 g
D28525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D28521G Supplier TCI America Supplier No. D28521G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 25676-62-4
Molecular Formula C7H16S4
Molecular Weight (g/mol) 228.45
MDL Number MFCD00506918
InChI Key XYHKMRNYSMEEDT-UHFFFAOYSA-N
PubChem CID 373740
IUPAC Name 2-({3-[(2-sulfanylethyl)sulfanyl]propyl}sulfanyl)ethane-1-thiol
SMILES SCCSCCCSCCS

Specifications

Color Yellow
Quantity 1 g
Synonym 1,3-Bis(2-mercaptoethylthio)propane, 2,2′C-(Trimethylenedithio)diethanethiol
Formula Weight 228.45
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3,7-Dithia-1,9-nonanedithiol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.