Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-(7-(Tert-Butoxycarbonyl)-5 6 7 8-Tetrahydroimidazo[1 2-A]Pyrazin-3-Yl)Propanoic Acid

Catalog No. 92050950MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
92-050-950MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 92-050-950MG Supplier Sigma Aldrich Supplier No. 92050950MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3-(7-(tert-Butoxycarbonyl)-5,6,7,8-tetrahydroimidazo[1,2-a]pyrazin-3-yl)propanoic acid is useful as a rigid linker in PROTAC development for targeted protein degradation as well as chemical conjugates. Incorporation of rigidity into the linker region of bifunctional protein degraders may impact the 3D orientation of the degrader and thus ternary complex formation as well as optimization of drug-like properties. Developed in partnership with ComInnex.Similar linkers in this series:920568920657ENAH38A2CCD6AC5372910424Portal: Building PROTAC™ Degraders for Targeted Protein DegradationCrosslinkers for BioconjugationTechnology Spotlight: Degrader Building Blocks for Targeted Protein DegradationPROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

Specifications

CAS 21149-17-7
Linear Formula C6H5CH2O2CNHC(=CH2)CO2CH3
Quantity 50 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.