Learn More
MSC Made in North America 88548268 3/8-16 UNC, 1" Length Under Head Hex Head Cap Screw Fully Threaded, Grade 5 Steel, Zinc-Plated Finish, 9/16" Hex
Catalog No. NC1395058
Encompass
Made in North America 88548268 3/8-16 UNC, 1" Length Under Head Hex Head Cap Screw Fully Threaded, Grade 5 Steel, Zinc-Plated Finish, 9/16" Hex
Catalog No. NC1395058 Supplier MSC Supplier No. 88548268
Made in North America 88548268 3/8-16 UNC, 1" Length Under Head Hex Head Cap Screw Fully Threaded, Grade 5 Steel, Zinc-Plated Finish, 9/16" Hex