Learn More
Ace Glass, Inc. 3/8" ID tubing hose connection on 5mm OD tube with 11mm OD second ring
SDP
Supplier:  Ace Glass, Inc. 846912
Encompass_Preferred
3/8 I.D. tubing hose connection on 5mm O.D. tube, with 11mm O.D. second ring. Four ring connector is tooled with flare and exposed for direct sealing to apparatus. Approx. 5-6 between connectors.
Catalog No. 50-974-611