Learn More
Chemglass Life Sciences Adapter, Flexible, Cajon Ultra-Torr, Use with 3/8in OD Tube, 6in Flexible Length, Airfree, Schlenk
SDP
Catalog No. AF006612
Encompass_Preferred
Cajon adapters provide the flexibility of rubber tubing and are the ideal replacement for rubber, plastic, or glass tubing in critical vacuum applications. Connections compensate for misalignment, expansion and contraction; they are compressible by at least 20%, and they are extendable by 50% of its nominal listed length. Cajon adapters are supplied complete with two AF-0067 unions and listed length of flexible tubing. PLEASE NOTE: SINCE FLEXIBLE TUBING WILL CONTRACT WHEN EVACUATED, GLASS APPARATUS SHOULD NOT BE FIRMLY CLAMPED IN PLACE UNTIL VACUUM IS ESTABLISHED WITHIN THE SYSTEM.
Catalog No. AF006612 Supplier Chemglass Life Sciences Supplier No. AF006612
Cajon adapters provide the flexibility of rubber tubing and are the ideal replacement for rubber, plastic, or glass tubing in critical vacuum applications. Connections compensate for misalignment, expansion and contraction; they are compressible by at least 20%, and they are extendable by 50% of its nominal listed length. Cajon adapters are supplied complete with two AF-0067 unions and listed length of flexible tubing. PLEASE NOTE: SINCE FLEXIBLE TUBING WILL CONTRACT WHEN EVACUATED, GLASS APPARATUS SHOULD NOT BE FIRMLY CLAMPED IN PLACE UNTIL VACUUM IS ESTABLISHED WITHIN THE SYSTEM.