Learn More
Bellco Glass, Inc. 3/8 Addition Port Assembly For 3,7 & 10 Bioreactor System
SDP
Catalog No. 501944719
Encompass_Preferred
3/8" addition port assembly for 3, 7 and 10 liter bioreactor systems/vessels. MADE IN THE USA English threads Fits .375" hole in headplate Dimensions: 2" oal Tube: 1/4" od x 5/32' id Material: SST 316l - ra 32 finish - inside & out With hose barb
Catalog No. 50-194-4719 Supplier Bellco Glass, Inc. Supplier No. 196402010
3/8" addition port assembly for 3, 7 and 10 liter bioreactor systems/vessels. MADE IN THE USA English threads Fits .375" hole in headplate Dimensions: 2" oal Tube: 1/4" od x 5/32' id Material: SST 316l - ra 32 finish - inside & out With hose barb