Learn More
Medchemexpress LLC 3,8-Diamino-6-phenylphenanthridine | 52009-64-0 | 285.34 | 1 G
SDP
Catalog No. 5000455097
Encompass_Preferred
3,8-Diamino-6-phenylphenanthridine is a biochemical assay reagent. It can bind to DNA by means of a non-specific binding along the DNA exterior.
  • Can bind to DNA by means of a non-specific binding along the DNA exterior.
  • Biochemical assay reagent.
Catalog No. 50-004-55097 Supplier Medchemexpress LLC Supplier No. HYW0115111G
3,8-Diamino-6-phenylphenanthridine is a biochemical assay reagent. It can bind to DNA by means of a non-specific binding along the DNA exterior.
  • Can bind to DNA by means of a non-specific binding along the DNA exterior.
  • Biochemical assay reagent.