Learn More
Medchemexpress LLC 3,8-Diamino-6-phenylphenanthridine | 52009-64-0 | 285.34 | 5 G
SDP
Catalog No. 5000455101
Encompass_Preferred
3,8-Diamino-6-phenylphenanthridine can bind to DNA by means of a non-specific binding along the DNA exterior. This product is intended for research use only and has not been fully validated for medical applications.
  • Binds to DNA non-specifically.
  • Acts as a biochemical assay reagent.
Catalog No. 50-004-55101 Supplier Medchemexpress LLC Supplier No. HYW0115115G
3,8-Diamino-6-phenylphenanthridine can bind to DNA by means of a non-specific binding along the DNA exterior. This product is intended for research use only and has not been fully validated for medical applications.
  • Binds to DNA non-specifically.
  • Acts as a biochemical assay reagent.